4-Acetamidobenzaldehyd 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-(4-Formylphenyl)acetamide, 4'-Formylacetanilide

400020100 400020250 400021000
ACRO400020100EA 69.5 EUR
ACRO400020100 ACRO400020250 ACRO400021000
4-Acetamidobenzaldehyd 98%
4-Acetamidobenzaldehyd
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Schmelzpunkt: 154…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003380
CAS-Nummer: 122-85-0
EINECS: 204-579-2

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 154 to 158 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR