9-Acridincarbonsäurehydrat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Acridine-9-carboxylic acid xhydrate, 9-Acridinecarboxylic Acid Hydrate, Acridine-9-carboxylic acid hydrate

400220050 400220010 400220250
ACRO400220050EA 205 EUR
ACRO400220050 ACRO400220010 ACRO400220250
9-Acridincarbonsäurehydrat 97%
9-Acridincarbonsäurehydrat

CAS No.: 332927-03-4

Formel: C₁₄H₁₁NO₃
Molecular Weight: 241.24 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149578
CAS-Nummer: 332927-03-4

Bestellen

Spezifikation Testergebnisse

Appearance Bright yellow to yellow-green Crystalline powder
Infrared spectrum Conforms
Water ≤15 % (K.F.)
Titration with NaOH ≥96.0 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR