DL-Alanine ethyl ester hydrochloride 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 2-aminopropanoate hydrochloride, H-DL-Ala-OEt.HCl

400430250
ACRO400430250EA 133 EUR
ACRO400430250
DL-Alanine ethyl ester hydrochloride 99%
DL-Alanine ethyl ester hydrochloride

Appearance: White Powder and/or granules

Formel: C₅H₁₂ClNO₂
Molecular Weight: 153,606 g/mol
Schmelzpunkt: 87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013018
CAS-Nummer: 617-27-6

Bestellen

Spezifikation Testergebnisse

Appearance White Powder and/or granules
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR