5-Aminofluorescein (isomer I), rein

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one (isomer I), 5-Amino-3',6'-dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 5-Aminofluorescein, Fluoresceinamine, isomer I, 5-Amino-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one, Fluoresceinamine (isomer I)

400770010 400770050
ACRO400770010EA 107 EUR
ACRO400770010 ACRO400770050
5-Aminofluorescein (isomer I), rein
5-Aminofluorescein (isomer I)
Formel: C₂₀H₁₃NO₅
Molecular Weight: 347.33 g/mol
Schmelzpunkt: 223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005052
CAS-Nummer: 3326-34-9

Bestellen

Spezifikation Testergebnisse

Appearance Dark red-brown Crystalline powder
Infrared spectrum Conforms
HPLC 95 % Typical
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR