3,4-Dicyananilin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
4-Aminophthalonitril, 4-Aminophthalsäuredinitril
401020100
401020250
401020050
ACRO401020100EA
287
EUR
ACRO401020100
ACRO401020250
ACRO401020050
3,4-Dicyananilin 97%
3,4-Dicyananilin
Formel:
C₈H₅N₃ Molecular Weight: 143.15 g/mol Siedepunkt: 321 Schmelzpunkt: 177…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041741 CAS-Nummer: 56765-79-8 EINECS: 260-370-6 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to yellow to orange Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 177 to 182 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...