Ammoniumcarbonat ACS

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ammoniumcarbonat, Hirschhornsalz

401130010 401130250 401130025
ACRO401130010EA 124 EUR
ACRO401130010 ACRO401130250 ACRO401130025
Ammoniumcarbonat ACS
Ammoniumcarbonat (enthält Ammoniumcarbamat)
Formel: (NH₄)₂CO₃
Molecular Weight: 96.09 g/mol
Schmelzpunkt: 58 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00010890
CAS-Nummer: 506-87-6
EINECS: 208-058-0
Merck Index: 13,00508

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or crystals and/or chunks
Non-volatile matter ≤0.01 %
Titration excess HCl ≥30.0 % (NH3)
Chloride (Cl) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Sulfur compounds (as SO4) ≤0.002 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR