Aluminon ACS

Lieferant: Thermo Fisher Scientific
Achtung

401500250 401501000
ACRO401500250EA 53 EUR
ACRO401500250 ACRO401501000
Aluminon ACS
Aluminon
Formel: C₂₂H₂₃N₃O₉
Schmelzpunkt: 220…225 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040925
CAS-Nummer: 569-58-4

Bestellen

Spezifikation Testergebnisse

Appearance Red to yellow-brown to red-brown Crystalline powder
Infrared spectrum Conforms
Residue after ignition ≤0.2 %
Sensitivity (to Aluminium) Passes test
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR