Sodium benzenesulphonate 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Benzenesulphonic acid Sodium Salt, Benzenesulphonic acid, sodium salt, Benzenesulphonate sodium
401850250
401851000
401855000
ACRO401850250EA
29.1
EUR
ACRO401850250
ACRO401851000
ACRO401855000
Sodium benzenesulphonate 98%
Sodium benzenesulphonate
CAS No.: 515-42-4
Formel:
C₆H₆O₃S Molecular Weight: 180.15 g/mol Schmelzpunkt: 450 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065179 CAS-Nummer: 515-42-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % (1 g, 105°C, 3 h) |
Solubility | (5% in water) Clear colorless to almost colorless |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...