Sodium benzenesulphonate 98%

Lieferant: Thermo Fisher Scientific

Synonyme: Benzenesulphonic acid Sodium Salt, Benzenesulphonic acid, sodium salt, Benzenesulphonate sodium

401850250 401851000 401855000
ACRO401850250EA 29.1 EUR
ACRO401850250 ACRO401851000 ACRO401855000
Sodium benzenesulphonate 98%
Sodium benzenesulphonate

CAS No.: 515-42-4

Formel: C₆H₆O₃S
Molecular Weight: 180.15 g/mol
Schmelzpunkt: 450 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065179
CAS-Nummer: 515-42-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (1 g, 105°C, 3 h)
Solubility (5% in water) Clear colorless to almost colorless
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR