Benzyldimethylhexadecylammoniumchlorid Hydrat 97%

Lieferant: Thermo Fisher Scientific
Gefahr

402070050 402075000
ACRO402070050EA 19.8 EUR
ACRO402070050 ACRO402075000
Benzyldimethylhexadecylammoniumchlorid Hydrat 97%
Benzyldimethylhexadecylammoniumchlorid Hydrat
Formel: C₂₅H₄₆ClN
Schmelzpunkt: 54…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149967
CAS-Nummer: 122-18-9
UN: 3261
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 % (on anhydrous substance)
Water ≤10 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR