4,4'-Bis(dimethylamino)benzil 98%
Lieferant: Thermo Scientific
Synonyme:
1,2-Bis(4-(dimethylamino)phenyl)ethane-1,2-dione, 1,2-Bis(4-dimethylaminophenyl)-1,2-ethanedione
402390010
ACRO402390010EA
43.4
EUR
ACRO402390010
4,4'-Bis(dimethylamino)benzil 98%
4,4'-Bis(dimethylamino)benzil
CAS No.: 17078-27-2
Formel:
C₁₈H₂₀N₂O₂ Molecular Weight: 296,37 g/mol Schmelzpunkt: 200…204 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051389 CAS-Nummer: 17078-27-2 |
Spezifikation Testergebnisse
Appearance | Yellow to green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 200 to 204 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...