Bis(2-hydroxyethyl)dimethylammonium chloride 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
402530250
ACRO402530250EA
98.8
EUR
ACRO402530250
Bis(2-hydroxyethyl)dimethylammonium chloride 99%
Bis(2-hydroxyethyl)dimethylammonium chloride
Appearance: White Crystalline powder
Formel:
C₆H₁₆ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041987 CAS-Nummer: 38402-02-7 |
Spezifikation Testergebnisse
Appearance | Colorless to white to off-white Adhering crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...