Bis(2,4,6-trichlorphenyl)oxalat 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Bis(2,4,6-trichlorophenyl) oxalate, TCPO, Oxalic Acid Bis(2,4,6-trichlorophenyl) Ester

402660050
ACRO402660050EA 140 EUR
ACRO402660050
Bis(2,4,6-trichlorphenyl)oxalat 98%
Bis(2,4,6-trichlorophenyl)oxalate

Appearance: White Crystalline powder

Formel: C₁₄H₄Cl₆O₄
Molecular Weight: 448,88 g/mol
Schmelzpunkt: 188…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043061
CAS-Nummer: 1165-91-9

Bestellen

Spezifikation Testergebnisse

Appearance White Powder or crystals
Infrared spectrum Conforms
Melting point 187 to 193 °C
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR