Bromthymolblau Natriumsalz ACS
Lieferant: Thermo Fisher Scientific
403260100
ACRO403260100EA
51.8
EUR
ACRO403260100
Bromthymolblau Natriumsalz ACS
Bromthymolblau Natriumsalz
Formel:
C₂₇H₂₇Br₂NaO₅S Schmelzpunkt: > 200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077263 CAS-Nummer: 34722-90-2 EINECS: 252-169-7 |
Spezifikation Testergebnisse
Appearance | Dark green to orange-brown Powder |
Infrared spectrum | Conforms |
Visual transition interval | from pH 6.0 (yellow) to pH 7.6 (blue) |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...