Calciumpropionat 98+%

Lieferant: Thermo Fisher Scientific

Synonyme: Propionsäure Calciumsalz, Calciumdipropionat

403900010 403900050 403900251
ACRO403900010EA 55.7 EUR
ACRO403900010 ACRO403900050 ACRO403900251
Calciumpropionat 98+%
Calciumpropionat
Formel: (CH₃CH₂COO)₂Ca
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036137
CAS-Nummer: 4075-81-4
EINECS: 223-795-8
Merck Index: 13,01703

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration Complexometric ≥98.0 % (on dry substance)
Loss on drying ≤4 % (105°C, 2 Hrs)
pH 6.9 to 9.0 (10 % solution)
Fluoride (F) ≤10 ppm
Arsenic (As) ≤3 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.3 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR