Cellulose acetate butyrate (35 - 39% butyryl)

Lieferant: Thermo Scientific

Synonyme: cellaburate

404255000
ACRO404255000EA 116 EUR
ACRO404255000
Cellulose acetate butyrate (35 - 39% butyryl)
Cellulose acetate butyrate

Appearance: White Crystalline powder

Formel: C₈₄H₁₃₀O₄₉
Molecular Weight: 1923,90 g/mol
Schmelzpunkt: 127…240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00081513
CAS-Nummer: 9004-36-8

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 195 to 205 °C
Viscosity 17 to 28 s
Average molecular weight 70000 (Mn)
Acetyl content 12 to 15 %
Hydroxyl content 1.8 % Typical
Butyryl content 35 to 39 %
Free acid (CH3COOH) ≤0.03 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR