Natriumcholat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Natrium-3alpha,7alpha,12alpha-trihydroxy-5beta-cholanoat, Natrium-3α,7α,12α-trihydroxy-5β-cholanoat, 3α,7α,12α-Trihydroxycholan-24-säure Natriumsalz, 3α,7α,12α-Trihydroxycholansäure Natriumsalz, 3α,7α,12α-Trihydroxy-5β-cholan-24-säure Natriumsalz, Cholsäure Natriumsalz, 3α,7α,12α-Trihydroxy-5β-cholansäure Natriumsalz

405191000 405195000
ACRO405191000EA 128 EUR
ACRO405191000 ACRO405195000
Natriumcholat 99%
Natriumcholat
Formel: C₂₄H₃₉NaO₅
Molecular Weight: 430.56 g/mol
Schmelzpunkt: 198
Lagertemperatur: Raumtemperatur
MDL: MFCD00150749
CAS-Nummer: 361-09-1
EINECS: 206-643-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5.0 % (110°C)
pH 7.5 to 9.5 (5 % in water)
Specific optical rotation +33° to +37° (20°C, 589 nm) (c=0.6, ethanol)
Titration with HClO4 98.0 to 102.0 % (On dried basis)
Heavy metals (as Pb) ≤50 ppm
Impurity ≤2.0 % Sodium deoxycholate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR