Kristallviolett, rein, zertifiziert

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cresyl echt violet, Cresyl violet, Cresyl fast violet, 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 9-Amino-5-imino-5H-benzo[a]phenoxazine acetate salt, Cresyl Violet acetate, Gentian violet

405760025 405760100 405760500
ACRO405760025EA 80.7 EUR
ACRO405760025 ACRO405760100 ACRO405760500
Kristallviolett, rein, zertifiziert
Cresylviolett (Azetat)
Formel: C₂₅H₃₀ClN₃
Molecular Weight: 321.33 g/mol
Schmelzpunkt: 140…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013151
CAS-Nummer: 548-62-9
EINECS: 208-953-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to black Crystalline powder
Infrared spectrum Conforms
Dye content ≥65 %
UV Lambda max 596 to 599 nm
Absorption Ratio P-15/P+15 0.94 to 1.2
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR