trans-1,4-Cyclohexanedimethanol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
trans-1,4-Bis(hydroxymethyl)cyclohexane
406062500
ACRO406062500EA
294
EUR
ACRO406062500
trans-1,4-Cyclohexanedimethanol 98%
trans-1,4-Cyclohexanedimethanol
Appearance: White Crystalline powder
Formel:
C₈H₁₆O₂ Molecular Weight: 144,21 g/mol Schmelzpunkt: 61…66 °C Dichte: 1,02 g/cm³ Flammpunkt: >109 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066360 CAS-Nummer: 3236-48-4 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 61 to 66 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...