1,1'-Diethyl-2,2'-cyanine Iodide 99%

Lieferant: Thermo Fisher Scientific
Gefahr

407255000
ACRO407255000EA 148 EUR
ACRO407255000
1,1'-Diethyl-2,2'-cyanine Iodide 99%
1,1'-Diethyl-2,2'-cyanine iodide

Appearance: Golden to dark red Crystalline powder

Formel: C₂₃H₂₃IN₂
Schmelzpunkt: 273 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011971
CAS-Nummer: 977-96-8
UN: 2811
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance Golden to dark red Crystalline powder
Infrared spectrum Conforms
UV spectrophotometry ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR