2-(Dimethylamino)ethyl-acetat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Acetic Acid 2-(Dimethylamino)ethyl Ester
408060250
ACRO408060250EA
77.5
EUR
ACRO408060250
2-(Dimethylamino)ethyl-acetat 98%
2-(Dimethylamino)ethyl acetate
Appearance: Clear light yellow Liquid
Formel:
C₆H₁₃NO₂ Molecular Weight: 131,18 g/mol Siedepunkt: 152…154 °C Dichte: 0,92 g/cm³ Flammpunkt: 47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041912 CAS-Nummer: 1421-89-2 UN: 1992 ADR: 3,III |
Spezifikation Testergebnisse
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4155 to 1.4175 (20°C, 589 nm) |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...