Dimethylglyoxim 99+% ACS

Lieferant: Thermo Fisher Scientific
Gefahr

408330250 408331000 408335000 408330025
ACRO408330250EA 39.7 EUR
ACRO408330250 ACRO408331000 ACRO408335000 ACRO408330025
Dimethylglyoxim 99+% ACS
Dimethylglyoxim
Formel: CH₃C(=NOH)C(=NOH)CH₃
Siedepunkt: 53 °C (8 mmHg)
Schmelzpunkt: 239…242 °C
Dichte: 1,37 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002117
CAS-Nummer: 95-45-4
EINECS: 202-420-1
UN: 2926
ADR: 4.1,III
Merck Index: 13,03272

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 239 to 242 °C (Dec)
Residue after ignition ≤0.05 %
Gravimetry ≥99.0 %
Suitability (for Ni determination) Passes test
Insoluble matter ≤0.05 % (in ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR