3,4-Dinitrobenzoesäure 99%

Lieferant: Thermo Scientific
Achtung

408700050
ACRO408700050EA 54.1 EUR
ACRO408700050
3,4-Dinitrobenzoesäure 99%
3,4-Dinitrobenzoic acid
Formel: C₇H₄N₂O₆
Molecular Weight: 212,12 g/mol
Schmelzpunkt: 163…166 °C
Dichte: 1,674 g/cm³ (20 °C)
MDL: MFCD00007255
CAS-Nummer: 528-45-0
EINECS: 208-433-9
Merck Index: 13,03303

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 163 to 166 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR