trans,trans-Dibenzylidenaceton 98%

Lieferant: Thermo Fisher Scientific

Synonyme: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

409000250 409001000 409000050
ACRO409000250EA 87.8 EUR
ACRO409000250 ACRO409001000 ACRO409000050
trans,trans-Dibenzylidenaceton 98%
trans,trans-Dibenzylidenaceton
Formel: C₁₇H₁₄O
Molecular Weight: 234.30 g/mol
Schmelzpunkt: 107…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004790
CAS-Nummer: 35225-79-7

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 108 to 114 °C
Assay ≥97.5 %
Pore volume 0.80 to 1.00 ml/g
Carbon (C) ≥16 %
Volatile matter ≤4 % (110 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR