Dodecylamine hydrochloride 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Dodecan-1-amine hydrochloride, Laurylamine Hydrochloride

409250100
ACRO409250100EA 55 EUR
ACRO409250100
Dodecylamine hydrochloride 99%
Dodecylamine hydrochloride

Appearance: White Powder

Formel: C₁₂H₂₇N
Molecular Weight: 221,81 g/mol
Schmelzpunkt: 186…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042017
CAS-Nummer: 929-73-7

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR