4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride Gemisch aus Mono- und Dihydrochlorid 98%

Lieferant: Thermo Scientific
Achtung

409680100
ACRO409680100EA 141 EUR
ACRO409680100
4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride Gemisch aus Mono- und Dihydrochlorid 98%
4-(4-Ethoxyphenylazo)-m-phenylenediamine, mixture of mono and dihydrochloride

Appearance: Dark brown to black Powder

Formel: C₁₄H₁₆N₄O·xHCl
Schmelzpunkt: 215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042023
CAS-Nummer: 74367-87-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark brown to black Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR