Ethyl-N-methylcarbamate 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-Methylurethane, N-Methylcarbamic Acid Ethyl Ester, Ethyl methylcarbamate

410260050
ACRO410260050EA 54.8 EUR
ACRO410260050
Ethyl-N-methylcarbamate 99%
Ethyl-N-methylcarbamate

Appearance: Clear colorless to light yellow-pink Liquid

Formel: C₄H₉NO₂
Molecular Weight: 103,12 g/mol
Siedepunkt: 170 °C (760 mmHg)
Dichte: 1,01 g/cm³
Flammpunkt: 61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041924
CAS-Nummer: 105-40-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow-pink Liquid
Infrared spectrum Conforms
Refractive index 1.4180 to 1.4200 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR