Ethylviolett 80%
Lieferant: Thermo Fisher Scientific
410490250
ACRO410490250EA
32.4
EUR
ACRO410490250
Ethylviolett 80%
Ethyl violet
Formel:
C₃₁H₄₂ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011765 CAS-Nummer: 2390-59-2 |
Spezifikation Testergebnisse
Appearance | Green Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤4 % |
Dye content | ≥75 % |
UV Lambda max | 593 to 599 nm (c=0.1, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...