Calcein

Lieferant: Thermo Fisher Scientific
Achtung

410630050 410630250
ACRO410630050EA 125 EUR
ACRO410630050 ACRO410630250
Calcein
Calcein
Formel: C₃₀H₂₆N₂O₁₃
Schmelzpunkt: 200 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005049
CAS-Nummer: 1461-15-0
EINECS: 215-957-1

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange-brown Powder
Infrared spectrum Conforms
UV spectrum Complies
Suitability (for Ca determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR