p-Hydrazinobenzenesulphonic acid hemihydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

411780500 411781000
ACRO411780500EA 82.2 EUR
ACRO411780500 ACRO411781000
p-Hydrazinobenzenesulphonic acid hemihydrate 98%
p-Hydrazinobenzenesulphonic acid hemihydrate

Appearance: Light yellow to light orange Powder

Formel: C₆H₈N₂O₃S
Schmelzpunkt: 285 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00480958
CAS-Nummer: 854689-07-9

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to light orange Powder
Infrared spectrum Conforms
Loss on drying 4 % to 5 %
Titration with NaOH ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR