2-Hydroxyacetophenon 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Phenacylhydroxyid, α-Hydroxyacetophenon, ω-Hydroxyacetophenon

411900250
ACRO411900250EA 140 EUR
ACRO411900250
2-Hydroxyacetophenon 97%
2-Hydroxyacetophenon
Formel: C₈H₈O₂
Molecular Weight: 136,15 g/mol
Siedepunkt: 118…120 °C (11 mmHg)
Schmelzpunkt: 83…88 °C
Dichte: 1,0963 g/cm³ (99 °C)
MDL: MFCD00041829
CAS-Nummer: 582-24-1
EINECS: 209-480-8

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow to orange Powder
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR