(S)-(-)-3-Hydroxy-3-phenylpropionsäure 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-3-Hydroxy-3-phenylpropanoic acid
412120010
ACRO412120010EA
105
EUR
ACRO412120010
(S)-(-)-3-Hydroxy-3-phenylpropionsäure 99%
(S)-(-)-3-Hydroxy-3-phenylpropionsäure
Formel:
C₉H₁₀O₃ Schmelzpunkt: 116…119 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00145262 CAS-Nummer: 36567-72-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystals or needles |
Infrared spectrum | Conforms |
Melting point | 116 to 119 °C |
Specific optical rotation | -21.5° to -23.5° (20 °C, 589 nm) (c=4, CH3OH) |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...