4-Iodbenzoylchlorid 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
412390250
412390050
ACRO412390250EA
144
EUR
ACRO412390250
ACRO412390050
4-Iodbenzoylchlorid 98%
4-Iodbenzoylchlorid
Formel:
C₇H₄ClIO Siedepunkt: 100 °C (0,7 mmHg) Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001058 CAS-Nummer: 1711-02-0 EINECS: 216-974-7 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | Light yellow to beige to light brown Crystalline powder and/or chunks or solid |
Infrared spectrum | Conforms |
Melting point | 62 to 66 °C |
GC | ≥97.5 % |
Acidity | ≤0.0002 meq/g |
Peroxides | ≤0.002 % (as H2O2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...