Sulphorhodamine B acid chloride, rein

Lieferant: Thermo Fisher Scientific
Achtung

413230010 413230050
ACRO413230010EA 188 EUR
ACRO413230010 ACRO413230050
Sulphorhodamine B acid chloride, rein
Sulphorhodamine B acid chloride

Appearance: Dark green Crystalline powder

Formel: C₂₇H₂₉ClN₂O₆S₂
Lagertemperatur: Kühlschrank
MDL: MFCD00042001
CAS-Nummer: 62796-29-6

Bestellen

Spezifikation Testergebnisse

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV Lambda max 561 to 567 nm (in CH2Cl2)
Molar extinction coefficient ≥85000 l/(mol.cm) (lambda max)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR