L(+)-Lysin Dihydrochlorid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Lys-OH.2HCl, (S)-(+)-2,6-Diaminohexansäure Dihydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Dihydrochlorid, (S)-(+)-Lysin Dihydrochlorid

413360250 413361000
ACRO413360250EA 31.2 EUR
ACRO413360250 ACRO413361000
L(+)-Lysin Dihydrochlorid 99%
L(+)-Lysin Dihydrochlorid
Formel: C₆H₁₆Cl₂N₂O₂
Molecular Weight: 219.11 g/mol
Schmelzpunkt: 202 °C
MDL: MFCD00066389
CAS-Nummer: 657-26-1
EINECS: 211-518-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 3 h)
Specific optical rotation +14° to +18° (20°C, 589 nm) (c=2, H2O)
Titration with HClO4 98.5 to 101.5 % (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR