DL-Lysin Monohydrochlorid 99%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-Lysin Hydrochlorid, (±)-Lysin Monohydrochlorid, (±)-2,6-Diaminohexansäure Hydrochlorid, (±)-2,6-Diaminocapronsäure Monohydrochlorid, (±)-2,6-Diaminohexansäure Monohydrochlorid, DL-Lysin Hydrochlorid, H-DL-Lys-OH.HCl, (±)-2,6-Diaminocapronsäure Hydrochlorid

413370250 413371000 413370010
ACRO413370250EA 40.3 EUR
ACRO413370250 ACRO413371000 ACRO413370010
DL-Lysin Monohydrochlorid 99%
DL-Lysin Monohydrochlorid
Formel: C₆H₁₅ClN₂O₂
Schmelzpunkt: 267 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064563
CAS-Nummer: 70-53-1
EINECS: 200-739-0

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation -1° to +1° (20°C, 589 nm) (c=1, water)
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR