Methyldiphenylamin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
414210050
414210250
ACRO414210050EA
42.8
EUR
ACRO414210050
ACRO414210250
Methyldiphenylamin 97%
Methyldiphenylamin
Formel:
C₁₃H₁₃N Schmelzpunkt: –7,6 °C Flammpunkt: >93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041900 CAS-Nummer: 552-82-9 EINECS: 209-023-2 |
Spezifikation Testergebnisse
Appearance | Clear yellow to orange-red Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6215 to 1.6235 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...