(R)-4-Phenylbutan-2-amine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (R)-(-)-1-Methyl-3-phenylpropylamine

414760010 414760050
ACRO414760010EA 152 EUR
ACRO414760010 ACRO414760050
(R)-4-Phenylbutan-2-amine 98%
(R)-4-Phenylbutan-2-amine

Appearance: Clear colorless Liquid

Formel: C₁₀H₁₅N
Dichte: 0,936 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00145208
CAS-Nummer: 937-52-0
UN: 2922
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5130 to 1.5150 (20°C, 589 nm)
Specific optical rotation -10° to -12° (20°C, 589 nm) (neat)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR