Naphthol AS-BI-phosphat 93%

Lieferant: Thermo Fisher Scientific
Achtung

415310010 415311000
ACRO415310010EA 332 EUR
ACRO415310010 ACRO415311000
Naphthol AS-BI-phosphat 93%
Naphthol AS-BI-phosphat
Formel: C₁₈H₁₅BrNO₆P
Lagertemperatur: Kühlschrank
MDL: MFCD00004058
CAS-Nummer: 1919-91-1
EINECS: 217-645-0

Bestellen

Spezifikation Testergebnisse

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥91 %
GC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR