New methylene Blue N, rein

Lieferant: Thermo Scientific

Synonyme: Basic Blue 24, Methylene Blue N

415530025
ACRO415530025EA 85.7 EUR
ACRO415530025
New methylene Blue N, rein
New methylene Blue N

Appearance: Green to dark brown Crystalline powder or crystals

Formel: C₁₈H₂₂ClN₃S
Molecular Weight: 347,91 g/mol
Schmelzpunkt: 190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135878
CAS-Nummer: 1934-16-3

Bestellen

Spezifikation Testergebnisse

Appearance Green to dark brown Crystalline powder or crystals
Infrared spectrum Conforms
Dye content ca 70%
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR