Nile blue A, rein, zertifiziert

Lieferant: Thermo Fisher Scientific

Synonyme: Nile blue A sulphate, Nile blue sulphate, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium sulphate, Basic Blue 12

415690100 415690500
ACRO415690100EA 37.2 EUR
ACRO415690100 ACRO415690500
Nile blue A, rein, zertifiziert
Nile blue A

CAS No.: 3625-57-8

Formel: C₄₀H₄₀N₆O₆S
Lagertemperatur: Raumtemperatur
MDL: MFCD00064529
CAS-Nummer: 3625-57-8

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to black Powder
Infrared spectrum Conforms
Dye content ≥70 %
UV Lambda max 635 to 646 nm
Absorption Ratio P-15/P+15 1.01 to 1.16
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR