Nilrot 99%, rein

Lieferant: Thermo Fisher Scientific

Synonyme: 9-(Diethylamino)-5H-benzo[a]phenoxazin-5-one

415711000 415710010
ACRO415711000EA 134 EUR
ACRO415711000 ACRO415710010
Nilrot 99%, rein
Nilrot

CAS No.: 7385-67-3

Formel: C₂₀H₁₈N₂O₂
Schmelzpunkt: 203…205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011639
CAS-Nummer: 7385-67-3

Bestellen

Spezifikation Testergebnisse

Appearance Dark green to dark red to brown Powder
Infrared spectrum Conforms
UV spectrum Complies
UV spectrophotometry ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR