Olivenöl, rein, raffiniert

Lieferant: Thermo Fisher Scientific

416530250 416535000 416530025
ACRO416530250EA 28 EUR
ACRO416530250 ACRO416535000 ACRO416530025
Olivenöl, rein, raffiniert
Olive oil
Dichte: 0,9135 g/cm³
Flammpunkt: 225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00131764
CAS-Nummer: 8001-25-0
EINECS: 232-277-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow-green Oily liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4710 (20°C, 589 nm)
Water ≤0.1 % (Coulometric)
Absorbance ≤1.20 (1g/100ml cyclohexane) measured at the max. between 264nm and 276nm
Composition ≤0.5 % (delta 7-Stigmasterol)
Acid value ≤0.3 mg KOH/g
Peroxide value ≤10.0 meq O2/kg
Unsaponifiable matter ≤1.5 %
Sesame oil Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR