Olivenöl, rein, unverarbeitet

Lieferant: Thermo Fisher Scientific

416540250 416545000 416540025
ACRO416540250EA 27.6 EUR
ACRO416540250 ACRO416545000 ACRO416540025
Olivenöl, rein, unverarbeitet
Olive oil
Dichte: 0,9135 g/cm³
Flammpunkt: 225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00131764
CAS-Nummer: 8001-25-0
EINECS: 232-277-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow to yellow-green Oily liquid
Infrared spectrum Conforms
Refractive index 1.4670 to 1.4710 (20°C, 589 nm)
Water ≤0.1 %
Absorbance ≤0.2 (270nm, 1g/100ml cyclohexane)
Composition ≤0.4 % (Eicosenoic acid)
Acid value ≤2.0 mg KOH/g
Peroxide value ≤20.0 meq O2/kg
Unsaponifiable matter ≤1.5 %
Sesame oil Complies


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR