Orange G, pure_certified
Lieferant: Thermo Fisher Scientific
Synonyme:
Acid Orange 10, Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat, Orange G Natriumsalz, CI 16230
416550100
416551000
ACRO416550100EA
31.4
EUR
ACRO416550100
ACRO416551000
Orange G, pure_certified
Orange G
Formel:
C₁₆H₁₀N₂Na₂O₇S₂ Molecular Weight: 452.38 g/mol Schmelzpunkt: 141 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012457 CAS-Nummer: 1936-15-8 EINECS: 217-705-6 |
Spezifikation Testergebnisse
Appearance | Orange Powder |
Infrared spectrum | Conforms |
Dye content | ≥80 % |
UV Lambda max | 476 to 481 nm |
Absorption Ratio P-15/P+15 | 0.88 to 0.95 |
BSC Certification | Passes |
Microbial contamination | (Salmonella) Absent in 375g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...