Orange G, pure_certified

Lieferant: Thermo Fisher Scientific

Synonyme: Acid Orange 10, Dinatrium-7-hydroxy-8-phenylazonaphthalin-1,3-disulfonat, Orange G Natriumsalz, CI 16230

416550100 416551000
ACRO416550100EA 31.4 EUR
ACRO416550100 ACRO416551000
Orange G, pure_certified
Orange G
Formel: C₁₆H₁₀N₂Na₂O₇S₂
Molecular Weight: 452.38 g/mol
Schmelzpunkt: 141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012457
CAS-Nummer: 1936-15-8
EINECS: 217-705-6

Bestellen

Spezifikation Testergebnisse

Appearance Orange Powder
Infrared spectrum Conforms
Dye content ≥80 %
UV Lambda max 476 to 481 nm
Absorption Ratio P-15/P+15 0.88 to 0.95
BSC Certification Passes
Microbial contamination (Salmonella) Absent in 375g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR