Peanut oil

Lieferant: Thermo Fisher Scientific

416855000 416850025
ACRO416855000EA 88.1 EUR
ACRO416855000 ACRO416850025
Peanut oil
Peanut oil
Dichte: 0,915 g/cm³
Flammpunkt: 283 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00131805
CAS-Nummer: 8002-03-7

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Oily liquid
Infrared spectrum Conforms
Water ≤0.1 %
Composition of fatty acids 0.5 to 3.0 % (GC) Lignoceric acid
Acid value ≤0.5 mg KOH/g
Peroxide value ≤5 meq O2/kg
Unsaponifiable matter ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR