N,N,N',N',N''-Pentamethyldiethylentriamin 98+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
416950250
416951000
416955000
416950000
ACRO416950250EA
37.4
EUR
ACRO416950250
ACRO416951000
ACRO416955000
416950000.
N,N,N',N',N''-Pentamethyldiethylentriamin 98+%
N,N,N',N',N''-Pentamethyldiethylentriamin
Formel:
C₉H₂₃N₃ Molecular Weight: 173.30 g/mol Schmelzpunkt: 18 °C Flammpunkt: 72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014876 CAS-Nummer: 3030-47-5 EINECS: 221-201-1 UN: 2922 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4415 to 1.4435 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...