Phenolrot ACS

Lieferant: Thermo Fisher Scientific
Achtung

417240050 417240100 417241000
ACRO417240050EA 37.2 EUR
ACRO417240050 ACRO417240100 ACRO417241000
Phenolrot ACS
Phenolrot
Formel: C₁₉H₁₄O₅S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003552
CAS-Nummer: 143-74-8
EINECS: 205-609-7
Merck Index: 13,07329

Bestellen

Spezifikation Testergebnisse

Appearance Red Powder
Infrared spectrum Conforms
Visual transition interval From pH 6.8 (yellow) to pH 8.2 (red)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR