Phenolrot Natriumsalz ACS

Lieferant: Thermo Fisher Scientific
Achtung

417250100 417250050 417251000
ACRO417250100EA 47 EUR
ACRO417250100 ACRO417250050 ACRO417251000
Phenolrot Natriumsalz ACS
Phenolrot Natriumsalz
Formel: C₁₉H₁₃NaO₅S
Lagertemperatur: Raumtemperatur
MDL: MFCD00066901
CAS-Nummer: 34487-61-1
EINECS: 252-057-8
Merck Index: 12,07397

Bestellen

Spezifikation Testergebnisse

Appearance Dark red to red-brown Powder
Infrared spectrum Conforms
Visual transition interval From pH 6.8 (yellow) to pH 8.2 (red)
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR